Få mere at vide
Tocris Bioscience™ NFκB p50 Transcription Factor Assay
GREENER_CHOICE
Artikelnummer. 18798036
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
18798036 25 mg
1 options
Artikelnummer. 18798036 Leverandør Tocris Bioscience™ Leverandørnr. 7051/25
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Indeholder
  • Transcription Factor Assay Plate
  • TFA Buffer, 5x
  • Blocking Reagent
  • Blocking Reagent
  • Primary Antibody
  • Gt X Rb, HRP Secondary Antibody
  • NFkB Capture Probe
  • NFkB Specific Competitor Oligonucleotide
  • TFA Negative Control Probe
  • HeLa Whole Cell Extract, TNFalpha Treated
  • Substrate (TMB/E)
  • Stop Solution
The transcription factor NFkB is involved in the expression and regulation of a number of important cellular and physiological processes such as growth, development, apoptosis, immune and inflammatory response
Tasquinimod is a high affinity negative allosteric modulator of HDAC4 (Kd = 10 - 30 nM). Binds the regulatory Zn2+ binding domain of HDAC4. Suppresses hypoxia-induced decrease in histone acetylation in human prostate cancer cells in vitro. Also binds S100A9. Antiangiogenic. Inhibits endothelial sprouting in vitro and growth of prostate tumor xenografts in nude mice. Orally bioavailable.