Få mere at vide
Nevirapine EP Impurity B, Mikromol™
Artikelnummer. 30051796
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051796 100 mg
30051856 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051796 Leverandør Mikromol Leverandørnr. MM1146.02
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nevirapine EP Impurity B
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
CAS 287980-84-1
Analyt- eller komponentnavne 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
Forsendelsestilstand Room Temperature
Molekylær formel C12 H10 N4 O
Certificeringer/compliance ISO 17025
Synonym Nevirapine Anhydrous Imp. B (EP), Nevirapine USP Related Compound B, Nevirapine USP RC B, 4-Methyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 6H-Dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 5,11-dihydro-4-methyl-, BIRG 616 BS, Nevirapine Related Compound B, Nevirapine Impurity B
InChI formel InChI=1S/C12H10N4O/c1-7-4-6-14-11-9(7)15-12(17)8-3-2-5-13-10(8)16-11/h2-6H,1H3,(H,15,17)(H,13,14,16)
SMIL Cc1ccnc2Nc3ncccc3C(=O)Nc12
IUPAC navn 4-methyl-5,11-dihydrodipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
Molekylvægt (g/mol) 226.23
Mængde 100 mg
Formel vægt 226.0855 g/mol