Få mere at vide
Nevirapine EP Impurity A, Mikromol™
Artikelnummer. 30052514
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052514 25 mg
30050082 100 mg
30461316 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30052514 Leverandør Mikromol Leverandørnr. MM1146.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nevirapine EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one
CAS 133627-17-5
Analyt- eller komponentnavne 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one
Forsendelsestilstand Room Temperature
Molekylær formel C14 H14 N4 O
Certificeringer/compliance ISO 17025
Synonym Nevirapine Anhydrous Imp. A (EP), Nevirapine USP Related Compound A, Nevirapine USP RC A, 11-Ethyl-4-methyl-5,11-dihydro-6H-dipyrido [3,2-b:2',3'-e][1,4]diazepin-6-one, Nevirapine Related Compound A, Nevirapine Impurity A
InChI formel InChI=1S/C14H14N4O/c1-3-18-12-10(5-4-7-15-12)14(19)17-11-9(2)6-8-16-13(11)18/h4-8H,3H2,1-2H3,(H,17,19)
SMIL CCN1c2nccc(C)c2NC(=O)c3cccnc13
IUPAC navn 11-ethyl-4-methyl-5H-dipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
Molekylvægt (g/mol) 254.29
Mængde 25 mg
Formel vægt 254.1168 g/mol