Få mere at vide
Nevirapine-12-(n-proylidenyl), TRC
Artikelnummer. 30465205
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30465205 25 mg -
30464407 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30465205 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39128025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 284686-27-7
Molekylær formel C18 H18 N4 O
Molekylvægt (g/mol) 306.36
Synonym 4-(But-1-en-1-yl)-11-cyclopropyl-5H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6(11H)-one, 4-(1-Buten-1-yl)-11-cyclopropyl-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
IUPAC navn 4-[(E)-but-1-enyl]-11-cyclopropyl-5H-dipyrido[3,2-a:2',3'-d][1,4]diazepin-6-one
SMIL CC\C=C\c1ccnc2N(C3CC3)c4ncccc4C(=O)Nc12
Tekniske data
Kemisk navn eller materiale Nevirapine-12-(n-proylidenyl)
Mængde 25 mg
InChI formel InChI=1S/C18H18N4O/c1-2-3-5-12-9-11-20-17-15(12)21-18(23)14-6-4-10-19-16(14)22(17)13-7-8-13/h3-6,9-11,13H,2,7-8H2,1H3,(H,21,23)
Formel vægt 306.1481