missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Neuropilin-1 Overexpression Lysate
Shop alle Bio Techne produkter
Klik for at se tilgængelige muligheder
Mængde:
0.1 mg
Beskrivelse
The Neuropilin-1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Tekniske data
Tekniske data
| Cellelinje | The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_1198. The protein contains a C-terminal DDK tag. |
| Til brug med (applikation) | Western Blot |
| Mængde | 0.1 mg |
| Opbevaringsbuffer | RIPA buffer |
| Renhed | Protein |
| Testspecificitet | Homo sapiens Neuropilin 1, mRNA. |
| Indhold og opbevaring | -80°C. Avoid freeze-thaw cycles. |
| Forskningskategori | Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Virology Bacteria and Parasites |
| Synonym | BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R |
| Proteintilstand | Native |
For Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion