missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Neuroligin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564404
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Immunogen sequence: PSLHPFGPFP PPPPTATSHN NTLPH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 88%.
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses.
Tekniske data
| Neuroligin 2 | |
| Polyclonal | |
| Unconjugated | |
| NLGN2 | |
| Kiaa1366; neuroligin 2; neuroligin-2; NL2; Nlgn2; ortholog of human and rat neuroligin 2 NLGN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57555 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFZ4 | |
| NLGN2 | |
| Recombinant protein corresponding to Human NLGN2. Recombinant protein control fragment (Product #RP-103833). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion