Få mere at vide
STEMCELL Technologies Neurodazine
Beskrivelse
- Neurodazine is a cell-permeable tri-substituted imidazole and neurogenic agent (Williams et al. 2007)
- Induces neurogenesis of C2C12 myoblasts as well as mature human muscle cells, marked by upregulation of neural genes (Williams et al. 2007; Williams et al. 2008)
- Induces neurogenesis and prevents astrocyte differentiation of P19 mouse embryonic carcinoma cells (Kim et al.)
- Induces neuronal differentiation of human (SH-SY5Y) and mouse (Neuro-2a) neuroblastoma cells, and mouse NIH3T3 fibroblast cells (Halder et al.)
Tekniske data
Tekniske data
| CAS | 937807-66-4 |
| Mængde | 10 mg |
| Molekylær formel | C27H21ClN2O3 |
| Renhed | ≥98% |