Få mere at vide
STEMCELL Technologies NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat)
Artikelnummer. 17108291
Encompass_Preferred
Skift visning
Mængde:
1 sæt
Artikelnummer. Mængde
17108291 1 sæt
1 options
Artikelnummer. 17108291 Leverandør STEMCELL Technologies Leverandørnr. 05715
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Kit for enzymatic dissociation of adult mouse and rat CNS tissue
  • NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat) is recommended for the enzymatic digestion and dissociation of adult mouse and rat central nervous system (CNS) tissue
  • NeuroCult™ Enzymatic Dissociation Kit has been optimized so that the entire procedure is reproducible, fast, and yields high cell numbers and viabilities
  • The resulting single-cell suspension is ready for immediate use in downstream applications
Tekniske data
Celletype Neural Stem and Progenitor Cells
Mængde 1 sæt