Få mere at vide
Netupitant N-Oxide, TRC
Artikelnummer. 30180230
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180230 25 mg
30180009 5 mg
30179782 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180230 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39095525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 910808-11-6
Molekylær formel C30H32F6N4O2
Molekylvægt (g/mol) 594.59
Synonym N,alpha,alpha-Trimethyl-N-[6-(4-methyl-4-oxido-1-piperazinyl)-4-(2-methylphenyl)-3-pyridinyl]-3,5-bis(trifluoromethyl)-benzeneacetamide
IUPAC navn 2-[3,5-bis(trifluoromethyl)phenyl]-N,2-dimethyl-N-[6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)-4-(2-methylphenyl)pyridin-3-yl]propanamide
SMIL CN(C(=O)C(C)(C)c1cc(cc(c1)C(F)(F)F)C(F)(F)F)c2cnc(cc2c3ccccc3C)N4CC[N+](C)([O-])CC4
Tekniske data
Kemisk navn eller materiale Netupitant N-Oxide
Mængde 25 mg
InChI formel InChI=1S/C30H32F6N4O2/c1-19-8-6-7-9-23(19)24-17-26(39-10-12-40(5,42)13-11-39)37-18-25(24)38(4)27(41)28(2,3)20-14-21(29(31,32)33)16-22(15-20)30(34,35)36/h6-9,14-18H,10-13H2,1-5H3
Formel vægt 594.2429