Få mere at vide
Nestin Antibody (196908), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1259MFV450
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Nestin Monoclonal antibody specifically detects Nestin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-reported
Tekniske data
| Nestin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cancer Stem Cells, Cell Biology, Cellular Markers, Extracellular Matrix, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 10763 | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 196908 | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-reported | |
| FLJ21841, nestin | |
| NS0 mouse myeloma cell line transfected with human Nestin | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |