missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nesprin-3 Rabbit anti-Human, Clone: S05-1B8, Novus Biologicals™
Rabbit Monoclonal Antibody
2215.00 DKK - 4475.00 DKK
Specifications
Antigen | Nesprin-3 |
---|---|
Clone | S05-1B8 |
Dilution | Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 ÎĽg/mL, Immunocytochemistry 5 ÎĽg/mL |
Applications | Western Blot, ELISA, Immunofluorescence, Immunocytochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310455
|
Bio-Techne
NBP3-15067-25UL |
25 ÎĽg |
2215.00 DKK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18341374
|
Bio-Techne
NBP3-15067-100UL |
100 ÎĽg |
4475.00 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nesprin-3 Monoclonal antibody specifically detects Nesprin-3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence, ImmunocytochemistrySpecifications
Nesprin-3 | |
Western Blot, ELISA 1:100-1:2000, Immunofluorescence 20 ÎĽg/mL, Immunocytochemistry 5 ÎĽg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
chromosome 14 open reading frame 49, FLJ25605, MGC75267, nesprin-3, NET53 | |
Recombinant protein of human Nesprin-3 (Uniprot#: Q6ZMZ3) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S05-1B8 | |
Western Blot, ELISA, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
161176 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nesprin-3 Rabbit anti-Human, Clone: S05-1B8, Novus Biologicals™