Få mere at vide
Nervonic Acid, TRC
Artikelnummer. 30132805
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132805 250 mg
30132454 1 g
30006571 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132805 Leverandør Toronto Research Chemicals Leverandørnr. TRCN390110250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 506-37-6
Molekylær formel C24 H46 O2
Molekylvægt (g/mol) 366.62
Synonym 15-Tetracosenoic acid, (15Z)- (9CI, ACI), (15Z)-15-Tetracosenoic acid (ACI), 15-Tetracosenoic acid, (Z)- (8CI), 15-Tetracosenoic acid, cis- (5CI), (15Z)-Tetracosenoic acid, (Z)-15-Tetracosenoic acid, 15Z-Tetracosenoic acid, cis-15-Tetracosenoic acid, Nervonic acid, Nevonic acid, Selacholeic acid
IUPAC navn (Z)-tetracos-15-enoic acid
SMIL CCCCCCCC\C=C/CCCCCCCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Nervonic Acid
Mængde 250 mg
InChI formel InChI=1S/C24H46O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h9-10H,2-8,11-23H2,1H3,(H,25,26)/b10-9-
Formel vægt 366.3498