Få mere at vide
Neratinib-d6, TRC
Artikelnummer. 30482403
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30482403 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482403 Leverandør Toronto Research Chemicals Leverandørnr. TRCN39009250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Neratinib-d6
CAS 1259519-18-0
Renhedsgrad noter HPLC
Molekylær formel C30H23D6ClN6O3
Mængde 50 mg
Synonym (2E)-N-[4-[[3-Chloro-4-(2-pyridinylmethoxy)phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino-d6)-2-butenamide, HKI 272-d6
InChI formel InChI=1S/C30H29ClN6O3/c1-4-39-28-16-25-23(15-26(28)36-29(38)9-7-13-37(2)3)30(20(17-32)18-34-25)35-21-10-11-27(24(31)14-21)40-19-22-8-5-6-12-33-22/h5-12,14-16,18H,4,13,19H2,1-3H3,(H,34,35)(H,36,38)/b9-7+/i2D3,3D3
SMIL O=C(/C=C/CN(C([2H])([2H])[2H])C([2H])([2H])[2H])NC1=CC2=C(C=C1OCC)N=CC(C#N)=C2NC3=CC(Cl)=C(OCC4=NC=CC=C4)C=C3
IUPAC navn (E)-4-(bis(methyl-d3)amino)-N-(4-((3-chloro-4-(pyridin-2-ylmethoxy)phenyl)amino)-3-cyano-7-ethoxyquinolin-6-yl)but-2-enamide
Molekylvægt (g/mol) 563.08
Formel vægt 562.2366
Procent renhed >95