Få mere at vide
Nepsilon,Nepsilon,Nepsilon-Trimethyllysine-d9, TRC
Artikelnummer. 30452127
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30452127 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452127 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79644725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Neta,Neta,Neta-Trimethyllysine-d9
CAS 1182037-78-0
Molekylær formel C9H11D9N2O2
Mængde 25 mg
Synonym (5S)-5-Amino-5-carboxy-N,N,N-tri(methyl-d3)-1-pentanaminium Inner Salt, Lysine betaine-d9, 6-N-Trimethyl L-Lysine-d9
SMIL [O-]C([C@@H](N)CCCC[N](C([2H])([2H+])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O
Molekylvægt (g/mol) 197.32
Formel vægt 197.209