Få mere at vide
NEpsilon-(1-Carboxymethyl)-L-lysine-d4, TRC
Artikelnummer. 30348947
Skift visning
Mængde:
0.5 mg
Artikelnummer. Mængde
30348947 0.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348947 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1800770.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Neta-(1-Carboxymethyl)-L-lysine-d4
CAS 936233-18-0
Renhedsgrad noter HPLC
Molekylær formel C8H12D4N2O4
Mængde 0.5 mg
Synonym 2-Amino-6-(carboxymethylamino)hexanoic Acid-d4, N6-(Carboxymethyl)lysine-d4, N6-(Carboxymethyl)-L-lysine-d4, CML-d4
InChI formel InChI=1S/C8H16N2O4/c9-6(8(13)14)3-1-2-4-10-5-7(11)12/h6,10H,1-5,9H2,(H,11,12)(H,13,14)/t6-/m0/s1/i1D2,2D2
SMIL N[C@H](C(O)=O)CC([2H])([2H])C([2H])([2H])CNCC(O)=O
IUPAC navn (2S)-2-amino-6-(carboxymethylamino)-4,4,5,5-tetradeuteriohexanoic acid
Molekylvægt (g/mol) 208.25
Procent renhed >95