Få mere at vide
NEpsilon-(1-Carboxyethyl)-L-lysine(Mixture of Diastereomers), TRC
Artikelnummer. 30011935
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011935 100 mg
30010417 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011935 Leverandør Toronto Research Chemicals Leverandørnr. TRCC178070100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5746-03-2
Molekylær formel C9H18N2O4
Molekylvægt (g/mol) 218.25
Synonym Nepsilon-(1-Carboxyethyl)lysine, N6-(1-Carboxyethyl)-L-lysine, N6-(1'-Carboxyethyl)lysine, CEL;
SMIL CC(NCCCC[C@H](N)C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Nepsilon-(1-Carboxyethyl)-L-lysine(Mixture of Diastereomers)
Renhedsgrad noter HPLC
Mængde 100 mg
Formel vægt 218.1267
Procent renhed >95%