Få mere at vide
Neoandrographolide, TRC
Artikelnummer. 30479417
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30479417 10 mg
30470181 5 mg
30474387 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30479417 Leverandør Toronto Research Chemicals Leverandørnr. TRCN38991310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27215-14-1
Molekylær formel C26 H40 O8
Molekylvægt (g/mol) 480.59
Synonym 2(5H)-Furanone, 3-[2-[(1R,4aS,5R,8aS)-5-[(β-D-glucopyranosyloxy)methyl]decahydro-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-, 2(5H)-Furanone, 3-[2-[5-[(β-D-glucopyranosyloxy)methyl]decahydro-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-, [1R-(1α,4aβ,5α,8aα)]-, 5β,9βH,10α-Labda-8(20),13-dien-16-oic acid, 18-(glucosyloxy)-15-hydroxy-, γ-lactone (8CI), 3-[2-[(1R,4aS,5R,8aS)-5-[(β-D-Glucopyranosyloxy)methyl]decahydro-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5H)-furanone, Neoandrographiside, Neoandrographolide
IUPAC navn 4-[2-[(1R,4aS,5R,8aS)-5,8a-dimethyl-2-methylidene-5-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]-3,4,4a,6,7,8-hexahydro-1H-naphthalen-1-yl]ethyl]-2H-furan-5-one
SMIL C[C@@]1(CO[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)CCC[C@@]3(C)[C@H](CCC4=CCOC4=O)C(=C)CC[C@H]13
Tekniske data
Kemisk navn eller materiale Neo-Andrographolide
Mængde 10 mg
InChI formel InChI=1S/C26H40O8/c1-15-5-8-19-25(2,14-33-24-22(30)21(29)20(28)18(13-27)34-24)10-4-11-26(19,3)17(15)7-6-16-9-12-32-23(16)31/h9,17-22,24,27-30H,1,4-8,10-14H2,2-3H3/t17-,18-,19-,20-,21+,22-,24-,25+,26+/m1/s1
Formel vægt 480.2723