Få mere at vide
Nemadipine B, TRC
Artikelnummer. 30483893
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483893 1 g
30478653 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483893 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3898001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79925-38-5
Molekylær formel C19 H21 Cl2 N O4
Molekylvægt (g/mol) 398.28
Synonym Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Felodipine Imp. C (EP)
IUPAC navn diethyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMIL CCOC(=O)C1=C(C)NC(=C(C1c2cccc(Cl)c2Cl)C(=O)OCC)C
Tekniske data
Kemisk navn eller materiale Diethyl 4-(2,3-Dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C19H21Cl2NO4/c1-5-25-18(23)14-10(3)22-11(4)15(19(24)26-6-2)16(14)12-8-7-9-13(20)17(12)21/h7-9,16,22H,5-6H2,1-4H3
Formel vægt 397.0848
Procent renhed >95%