missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
NELL2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00073IR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
NELL2 Polyclonal specifically detects NELL2 in Human samples. It is validated for ELISA.
Tekniske data
| NELL2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 755 | |
| nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 | |
| Produced in rabbits immunized with purified, recombinant NELL2 (Q99435; Met1-Leu816). | |
| 0.1 mL | |
| Neuroscience | |
| 4753 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion