Få mere at vide
Nelarabine, Tocris Bioscience™
GREENER_CHOICE
Purine nuceloside analog; inhibits DNA synthesis
Brand:  Tocris Bioscience 6359/50
Encompass
Produktkode 15963261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Nelarabine is a nucleoside analog. Metabolized by adenosine deaminase to active metabolite, arabinofuranosylguanine (ara-G). Inhibits DNA synthesis and induces apoptosis. Inhibits growth of human T cell lymphoblastic leukemia cells in vitro and in vivo.
Tekniske data
Tekniske data
9-β-D-Arabinofuranosyl-6-methoxy-9H-purin-2-amine
121032-29-9
>98%
50 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner