Få mere at vide
NEK7 Antibody [DyLight 755], Novus Biologicals Biologicals™
Artikelnummer. 30498963 Shop alle Bio Techne produkter
Skift visning
Mængde:
0.1 mL
Artikelnummer. Mængde
30498963 0.1 mL
1 options
Artikelnummer. 30498963 Leverandør Novus Biologicals Leverandørnr. NBP337907IR
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
NEK7 Polyclonal antibody specifically detects NEK7 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Tekniske data
Antigen NEK7
Applikationer ELISA, Western Blot
Klassifikation Polyclonal
Konjugeret DyLight 755
Formulering 50mM Sodium Borate
Gene Alias EC 2.7.11.1, Never in mitosis A-related kinase 7, NIMA (never in mitosis gene a)-related kinase 7, NimA-related protein kinase 7, serine/threonine-protein kinase Nek7
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 257-301 of human NEK7 (NP_598001.1).,, Sequence:, PLPSDHYSEELRQLVNMCINPDPEKRPDVTYVYDVAKRMHACTAS
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Protein Kinase
Primær eller sekundær Primary
Gen-id (Entrez) 140609
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG