Få mere at vide
(+)-Nefopam Hydrochloride, TRC
Artikelnummer. 30473110
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30473110 500 mg
30473243 250 mg
30478449 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30473110 Leverandør Toronto Research Chemicals Leverandørnr. TRCN385745500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 53648-31-0
Molekylær formel C17H19NO . HCl
Molekylvægt (g/mol) 289.8
Synonym (S)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, (1S)-3,4,5,6-Tetrahydro-5-methyl-1-phenyl-1H-2,5-benzoxazocine Hydrochloride, S-Nefopam Hydrochloride
IUPAC navn (1S)-5-methyl-1-phenyl-1,3,4,6-tetrahydro-2,5-benzoxazocine;hydrochloride
SMIL Cl.CN1CCO[C@@H](c2ccccc2)c3ccccc3C1
Tekniske data
Kemisk navn eller materiale (+)-Nefopam Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C17H19NO.ClH/c1-18-11-12-19-17(14-7-3-2-4-8-14)16-10-6-5-9-15(16)13-18;/h2-10,17H,11-13H2,1H3;1H/t17-;/m0./s1
Formel vægt 289.1233