missing translation for 'onlineSavingsMsg'
Få mere at vide

Nefopam hydrochloride, MedChemExpress

Artikelnummer. 16444537
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16444537 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16444537 Leverandør Medchem Express Leverandørnr. HYB1057

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Nefopam hydrochloride (Fenazoxine hydrochloride) is a centrally-acting but non-opioid analgesic drug, for the relief of moderate to severe pain. Nefopam hydrochloride targets β-catenin protein level in mesenchymal cells in-vitro and in-vivo.

Nefopam hydrochloride (Fenazoxine hydrochloride) is a centrally-acting but non-opioid analgesic drug, for the relief of moderate to severe pain. Nefopam hydrochloride targets β-catenin protein level in mesenchymal cells in-vitro and in-vivo.

Tekniske data

Kemisk navn eller materiale Nefopam hydrochloride
CAS 23327-57-3
Farve Off-White
Renhedsgrad noter Research
Mængde 50 mg
Synonym Fenazoxine hydrochloride
Opløselighedsinformation H2O : 12.5 mg/mL (43.13 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (43.13 mM; Need ultrasonic)
SMIL CN1CCOC(C2=CC=CC=C2)C3=CC=CC=C3C1.[H]Cl
Molekylvægt (g/mol) 289.8
Procent renhed 99.78%
Grad Research
Fysisk form Solid
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.