missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ NEFH Monoclonal Antibody (RMdO-20)

Artikelnummer. 10726653
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
200 μg
Pakningsstørrelse:
Hver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
10726653 200 μg Hver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10726653 Leverandør Invitrogen™ Leverandørnr. 131300

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

This antibody reacts with 200 kDa subunit protein of human neurofilament. It specifically recognizes a nonphosphorylated epitope in the tail domain of NF-H. Dephosphorylation of NF-H will increase immunoreactivity with this antibody. This antibody is suitable for immunohistochemical staining of Bouin's and alcohol-fixed paraffin-embedded or frozen tissue sections. To stain, incubate 30-60 minutes at room temperature or overnight at 4°C.

Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen NEFH
Applikationer ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon RMdO-20
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 0.1% sodium azide
Gene Nefh
Genadgangsnr. P12036, P16884, P19246
Gene Alias 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg
Gensymboler Nefh
Værtsarter Mouse
Immunogen Adult Rat neurofilaments.
Oprensningsmetode Purified
Mængde 200 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 100357177, 100753127, 101838568, 24587, 380684, 4744, 528842
Målarter Human, Mouse, Rat, Rabbit, Bovine, Hamster, Mollusca
Indhold og opbevaring -20°C
Produkttype Antibody
Form Liquid
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.