Få mere at vide
Neem Oil, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Neem Oil |
| CAS | 8002-65-1 |
| Mængde | 100 g |
| Synonym | Azadirachta indica oil, Azadirachta indica seed oil, Margosa oil, Margosa seed oil, Melia azadirachta oil, Melia azadirachta seed oil, Neem Indica, Neem seed oil |