Learn More
Trajan Scientific and Medical™ Replacement Needle for CTC PAL RTC LC Syringes
Reduce costs. Replace the needle not the syringe. For PAL RTC instruments.
Brand:  Trajan Scientific and Medical™ 037880
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13129035
  • 328.00 DKK / Pack of 2
Estimated Shipment: 27-05-2024
to see stock.
Description
Description
  • Increased application specific flexibility
  • Reduced overall costs by replacing the needle rather than the entire syringe
Specifications
Specifications
0.17 mm
57 mm
2/Pk.
CTC RTC & Thermo RSH Syringes (10μL)
0.72 mm
Gastight
LC
Fixed