Få mere at vide
(+)-Nebivolol Hydrochloride, TRC
Artikelnummer. 30483244
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30483244 0.5 mg 0.5mg
30480757 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483244 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3242400.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+)-Nebivolol Hydrochloride
CAS 213132-06-0
Molekylær formel C22H25F2NO4.HCl
Mængde 0.5 mg
Synonym (αR,α'R,2R,2'S)-α,α'-[Iminobis(methylene)]bis[6-fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol Hydrochloride (1:1) (9Cl), (R,R,R,S)-Nebivolol Hydrochloride
InChI formel InChI=1S/C22H25F2NO4.ClH/c23-15-3-7-19-13(9-15)1-5-21(28-19)17(26)11-25-12-18(27)22-6-2-14-10-16(24)4-8-20(14)29-22;/h3-4,7-10,17-18,21-22,25-27H,1-2,5-6,11-12H2;1H/t17-,18-,21-,22+;/m1./s1
SMIL O[C@@H]([C@]1([H])OC(C(CC1)=C2)=CC=C2F)CNC[C@H]([C@@](CCC3=C4)([H])OC3=CC=C4F)O.Cl
IUPAC navn (1R)-1-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-[[(2R)-2-[(2R)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]-2-hydroxyethyl]amino]ethanol;hydrochloride
Molekylvægt (g/mol) 441.9
Formel vægt 441.1518