Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UN160
Nearly gastight interior with possibility for inlet/outlet through two tubes with ball valves.
Brand: Memmert™ UN160K3
6819.32 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Gas inlet/outlet through 2 tubes with ball valves
- Technical clarification required for combination with other options
Specifications
UN160 Oven | |
Interior | |
Gas Inlet/Outlet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UN160 > For Use With: UN160
Spot an opportunity for improvement?Share a Content Correction