Beskrivelse
NDUFB10 Polyclonal antibody specifically detects NDUFB10 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Tekniske data
Tekniske data
| Antigen | NDUFB10 |
| Applikationer | ELISA, Immunoprecipitation, Western Blot |
| Klassifikation | Polyclonal |
| Konjugeret | DyLight 350 |
| Formulering | 50mM Sodium Borate |
| Gene Alias | CI-PDSW, complex I PDSW subunit, Complex I-PDSW, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10 (22kD, PDSW), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10, NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3), NADH-ubiquinone oxidoreductase PDSW subunit, PDSW |
| Værtsarter | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human NDUFB10 (NP_004539.1).,, Sequence:, MPDSWDKDVYPEPPRRTPVQPNPIVYMMKAFDLIVDRPVTLVREFIERQHAKNRYYYYHRQYRRVPDITECKEEDIMCMYEAEMQWKRDYKVDQEIINIMQDRLKACQQREGQNYQQNCIKEVEQFTQVAKAYQDRYQDLGAYSSARKCLAKQRQRMLQERKAAKEAAAATS |
| Oprensningsmetode | Affinity purified |
| Mængde | 0.1 mL |
| Vis mere |