missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ NCKIPSD Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15958334
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
Pakningsstørrelse:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15958334 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15958334 Leverandør Invitrogen™ Leverandørnr. PA565592

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Immunogen sequence: RKETLSRRGPS ASSVAVMTSS TSDHHLDAAA ARQPNGVCRA GFERQHSLPS SEHLGADGGL YQIPLPSSQI PPQP Highest antigen sequence identity to the following orthologs - mouse 72%, rat 69%.

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintenance of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen NCKIPSD
Applikationer Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 0.05 mg/mL
Konjugeret Unconjugated
Formulering PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCKIPSD
Genadgangsnr. Q9NZQ3
Gene Alias 54 kDa VacA-interacting protein; 54 kDa vimentin-interacting protein; 90 kDa N-WASP-interacting protein; 90 kDa SH3 protein interacting with Nck; AF3P21; dia interacting protein; dia-interacting protein 1; diaphanous protein interacting protein; Diaphanous protein-interacting protein; DIP; DIP1; DIP-1; mDia interacting protein-1; MGC23891; NCK interacting protein with SH3 domain; NCK-interacting protein with SH3 domain; Nckipsd; N-WASP-binding protein; ORF1; SH3 adapter protein SPIN90; SH3 protein interacting with Nck, 90 kDa; Spin90; VIP54; Wasbp; WASLBP; WASP-interacting SH3-domain protein; WISH; Wiskott-Aldrich syndrome homolog binding protein; Wiskott-Aldrich syndrome protein-binding protein; wiskott-Aldrich syndrome protein-binding protein WISH; wiskott-Aldrich syndrome protein-interacting protein
Gensymboler NCKIPSD
Værtsarter Rabbit
Immunogen Recombinant Human NCKIPSD. Recombinant protein control fragment (Product #RP-104887).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 51517
Målarter Human
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.