missing translation for 'onlineSavingsMsg'
Få mere at vide

NCGC00244536, MedChemExpress

Artikelnummer. 16496297
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16496297 5 mg 5mg
16479986 50 mg -
16464686 100 mg 100mg
16453578 10 mg -
16410909 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16496297 Leverandør Medchem Express Leverandørnr. HY101799

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress NCGC00244536 is a potent KDM4B inhibitor with an IC50 of 10 nM.

NCGC00244536 is a potent KDM4B inhibitor with an IC50 of 10 nM.

Specifications

Kemisk navn eller materiale NCGC00244536
CAS 2003260-55-5
Farve Yellowish Green
Renhedsgrad noter Research
Molekylær formel C25H22N2O2
Mængde 5 mg
Synonym KDM4B Inhibitor B3
Opløselighedsinformation DMSO : 36.67 mg/mL (95.88 mM; Need ultrasonic)
SMIL O=C(NCCCC1=CC=CC=C1)C2=CC=CC(C3=CC(O)=C4N=CC=CC4=C3)=C2
Molekylvægt (g/mol) 382.45
Formel vægt 382.45
Procent renhed 98.94%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.