Få mere at vide
Tocris™ NBC 6
Beskrivelse
NBC 6 is a NLRP3 inflammasome inhibitor (IC50 = 574 nM). Inhibits nigericin-induced inflammasome activation in THP-1 cells and Imiquimod (Cat. No. 3700) induced IL-1β release from LPS-primed bone marrow-derived macrophages (BMDMs).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (T4)-[2-Acetyl-4,4,4-trichloro-3-(imino-κN)butanamidito-κO]diphenylboron |
| CAS | 2068818-02-8 |
| Mængde | 10 mg |
| Mål | Inflammasomes Inhibitors |
| Molekylær formel | C18H16BCl3N2O2 |
| Formel vægt | 1869 |
| Renhed | 98% |