missing translation for 'onlineSavingsMsg'
Få mere at vide

Naxagolide-O-methyl Hydrochloride, TRC

Artikelnummer. 30481955
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30481955 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30481955 Leverandør Toronto Research Chemicals Leverandørnr. TRCN37880525MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Naxagolide-O-methyl Hydrochloride
CAS 88058-72-4
Molekylær formel C16H23NO2 . HCl
Mængde 25 mg
Synonym (4aR-trans)-3,4,4a,5,6,10b-Hexahydro-9-methoxy-4-propyl-2H-naphth[1,2-b]-1,4-oxazine Hydrochloride
InChI formel InChI=1S/C16H23NO2.ClH/c1-3-8-17-9-10-19-16-14-11-13(18-2)6-4-12(14)5-7-15(16)17;/h4,6,11,15-16H,3,5,7-10H2,1-2H3;1H/t15-,16-;/m1./s1
SMIL Cl.CCCN1CCO[C@H]2[C@H]1CCc3ccc(OC)cc23
IUPAC navn (4aR,10bR)-9-methoxy-4-propyl-2,3,4a,5,6,10b-hexahydrobenzo[h][1,4]benzoxazine;hydrochloride
Molekylvægt (g/mol) 297.82
Formel vægt 297.1496
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.