Få mere at vide
Tocris Bioscience™ NAV 2729
GREENER_CHOICE
Selective ARF6 inhibitor
Brand:  Tocris Bioscience™ 5986/50
Encompass
Produktkode 15717186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
NAV 2729 is a selective ARF6 inhibitor (IC50 = 1.0 μM). Selective for ARF6 over other ARF family members and small GTPases. Inhibits ARF6 activation in uveal melanoma cells in vitro and blocks downstream signaling pathways of oncogenic GNAQ. Reduces tumorigenesis in mouse model of uveal melanoma.
Tekniske data
Tekniske data
Yellow
419547-11-8
456.88g/mol
Store at 4°C
50 mg
ARF GTPase Inhibitors
C25H17ClN4O3
Soluble to 100mM in DMSO
Blocks downstream signaling pathways
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only