Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Description
- Only two operating points
- Evacuate via the new automatic valve
- Available in a range of colors
- One-handed design is the simplest safety pipette filler available
- Ability to clean inside of bulb by removing patented valve and rinsing out
Spécification
Spécification
| Color | Red |
| Material | Natural Rubber |
| Type | Safety Pipette Filler Flip |