Få mere at vide
Saint-Gobain Natural Rubber Folding Skirt Stoppers
Artikelnummer. 11362074
Skift visning
Farve:
Hvid
Pakningsstørrelse:
Hver
Artikelnummer. Farve unitSize
11362074 Hvid Hver
1 options
Artikelnummer. 11362074 Leverandør Saint-Gobain Leverandørnr. 407020
Keep container contents secure with these two-part folding skirt stoppers. Saint-Gobain Performance Plastics™ Natural Rubber Folding Skirt Stoppers are ideal for securing glass, metal and plastic containers.
  • Designed with a double seal
  • Upper hollow plug section can be perforated to extract contents without removing the stopper and remains watertight even after several perforations
  • The lower section grips the outside of the container and the upper hollow plug features annular serrations to provide a tight seal at the rim
  • Ideal for chemical transfer
  • Temperature resistant from -20 to +100°C
TRUSTED_SUSTAINABILITY
Tekniske data
Højde (metrisk) H1: 13.5mm; H2: 19mm
Størrelse 19.4
Farve Hvid
Diameter (metrisk) bund 15.8 mm
Diameter (metrisk) Top 17.8 mm
Materiale Rubber
Produkttype Folding Skirt Stopper