Få mere at vide
Abcam Native Staphylococcus aureus Protein A (HRP)
Artikelnummer. 32143602 Shop alle Abcam produkter
Skift visning
1 mg
Artikelnummer.
32143602 1 mg
1 options
Artikelnummer. 32143602 Leverandør Abcam Leverandørnr. ab74561mg
 Begrænset vare
Forventet forsendelse: 02-12-2026
Native Staphylococcus aureus Protein A (HRP) is a Staphylococcus aureus subsp. aureus NCTC 8325 Full Length protein, expressed in Native, with >95%, suitable for Dot, IM, WB, ELISA, IHC-P.
Native Staphylococcus aureus Protein A (HRP) is a Staphylococcus aureus subsp. aureus NCTC 8325 Full Length protein, expressed in Native, with >95%, suitable for Dot, IM, WB, ELISA, IHC-P.
The product is subject to the following: Abcam Restricted Use Statement