Få mere at vide
Thermo Scientific™ Nationale hætteglasknusere
Type:
Manuel Benchtop Vial Crusher
Pneumatisk bordmonteret hætteglasknuser
Pakningsstørrelse:
Hver
Beskrivelse
Reducer omkostningerne til bortskaffelse af affald ved at reducere den samlede mængde affald med Thermo Scientific™ National Vial Crusher.
- Breaks and separates liquid from all 1mL to 100mL vials and ampules
- Capable of processing approximately 5000 2mL vials per hour
- Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
- Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
Tekniske data
Tekniske data
| Oversigt | Pneumatic Benchtop Vial Crusher with Stainless-steel Mobile Cart |
| Inkluderer | Stainless Steel Mobile Cart |
| Produktlinje | National |
| Type | Pneumatisk bordmonteret hætteglasknuser |