Få mere at vide
Thermo Scientific™ National Scientific ASM Vials
Beskrivelse
- 100 vials and 100 caps with pre-assembled septa
- Reusable two-compartment trays protect vials and closures while keeping matching supplies together
- Clear trays make it easy to keep track of available supplies without opening containers
- Patched
Tekniske data
Tekniske data
| Lukningsfarve | Blue |
| Diameter (metrisk) åbning | 9 mm |
| Farve | Ravgul |
| Volumen (metrisk) | 2 mL |
| Septum | Bonded Clear PTFE/Clear Silicone |
| nr. pr. pakke | 100 |