Learn More
Thermo Scientific™ National Inserts for Standard-Opening Vials
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
Brand: Thermo Scientific™ C4013-K465
Additional Details : Weight : 0.34000kg
Description
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
- Pulled point inserts are an economical choice for non-critical applications
Specifications
C4013-K465 | |
250 μL | |
National |
Kimshield Glass | |
Flat Bottom | |
Insert |