Learn More
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC analyzers
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
Brand: Thermo Scientific™ C4011-1296
Additional Details : Weight : 8.37000kg
Description
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
- Maximum fill volume: 5mL
- Superior quality N-51A clear borosilicate glass
- 5mL sample vial is designed for use in the Shimadzu™ Model ASI-5000
- Screw thread vial fits standard 10-425 closures (included with pre-assembled product)
- Open-top vial was designed in cooperation with the original equipment manufacturer
Specifications
Open Top TOC Vial | |
5 mL | |
10-425 | |
Shimadzu Model ASI-5000 |