Få mere at vide
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC analyzers
Oversigt:
Open Top TOC Vial
Screw Thread TOC Vial, Polypropylene Cap with PTFE septum
Pakningsstørrelse:
Pakke med 100
Pakke med 1000
Beskrivelse
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
- Maximum fill volume: 5mL
- Superior quality N-51A clear borosilicate glass
- 5mL sample vial is designed for use in the Shimadzu™ Model ASI-5000
- Screw thread vial fits standard 10-425 closures (included with pre-assembled product)
- Open-top vial was designed in cooperation with the original equipment manufacturer
Tekniske data
Tekniske data
| Oversigt | Screw Thread TOC Vial, Polypropylene Cap with PTFE septum |
| Volumen (metrisk) Virker | 5 mL |
| Lukkestørrelse | 10-425 |
| Til brug med (udstyr) | Shimadzu Model ASI-5000 |