Få mere at vide
Nateglinide EP Impurity A, Mikromol™
Artikelnummer. 30052635
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30052635 25 mg
30049398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30052635 Leverandør Mikromol Leverandørnr. MM0847.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nateglinide EP Impurity A
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale trans-4-(1-methylethyl)cyclohexanecarboxylic acid
CAS 5-6-7077
Analyt- eller komponentnavne trans-4-(1-methylethyl)cyclohexanecarboxylic acid
Forsendelsestilstand Room Temperature
Molekylær formel C10 H18 O2
Certificeringer/compliance ISO 17025
Synonym trans-4-(1-Methylethyl)cyclohexanecarboxylic Acid, Nateglinide Imp. A (EP), Nateglinide USP Related Compound A, Nateglinide USP RC A, trans-4-(1-Methylethyl)cyclohexanecarboxylic Acid, Cyclohexanecarboxylic acid, 4-isopropyl-, trans- (8CI), trans-4-Isopropylcyclohexanecarboxylic acid, trans-p-Menthan-7-oic acid, Nateglinide Related Compound A, Nateglinide Impurity A
InChI formel InChI=1S/C10H18O2/c1-7(2)8-3-5-9(6-4-8)10(11)12/h7-9H,3-6H2,1-2H3,(H,11,12)/t8-,9-
SMIL CC(C)[C@@H]1CC[C@H](CC1)C(=O)O
IUPAC navn 4-propan-2-ylcyclohexane-1-carboxylic acid
Molekylvægt (g/mol) 170.25
Mængde 25 mg
Formel vægt 170.1307 g/mol