Få mere at vide
Nateglinide-d5, TRC
Artikelnummer. 30469020
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469020 10 mg
30472932 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469020 Leverandør Toronto Research Chemicals Leverandørnr. TRCN37937710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1227666-13-8
Molekylær formel C19 2H5 H22 N O3
Molekylvægt (g/mol) 322.45
Synonym D-Phenylalanine-2,3,4,5,6-d5, N-[[trans-4-(1-methylethyl)cyclohexyl]carbonyl]-, Nateglinide-D5, Nateglinide-D5 (Phenyl-D5), N-[[trans-4-(Propan-2-yl)cyclohexyl]carbonyl]-D-phenylalanine-d5, N-[[trans-4-(1-Methylethyl)cyclohexyl]carbonyl]-D-phenylalanine-2,3,4,5,6-d5
IUPAC navn (2R)-3-(2,3,4,5,6-pentadeuteriophenyl)-2-[(4-propan-2-ylcyclohexanecarbonyl)amino]propanoic acid
SMIL [2H]c1c([2H])c([2H])c(C[C@@H](NC(=O)[C@@H]2CC[C@H](CC2)C(C)C)C(=O)O)c([2H])c1[2H]
Tekniske data
Kemisk navn eller materiale Nateglinide phenyl-d5
Mængde 10 mg
InChI formel InChI=1S/C19H27NO3/c1-13(2)15-8-10-16(11-9-15)18(21)20-17(19(22)23)12-14-6-4-3-5-7-14/h3-7,13,15-17H,8-12H2,1-2H3,(H,20,21)(H,22,23)/t15-,16-,17-/m1/s1/i3D,4D,5D,6D,7D
Formel vægt 322.2305