Få mere at vide
Nardosinone, TRC
Artikelnummer. 30483682
Skift visning
Mængde:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483682 1 g
30479754 50 mg
30478485 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483682 Leverandør Toronto Research Chemicals Leverandørnr. TRCN3789301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23720-80-1
Molekylær formel C15H22O3
Molekylvægt (g/mol) 250.33
Synonym [3aR-(3aα,9α,9aα,9bα)]-1,3a,4,7,8,9,9a,9b-Octahydro-1,1,9,9a-tetramethyl-5H-Naphtho[2,1-c][1,2]dioxol-5-one, Nardosinon
IUPAC navn (3aR,9R,9aR,9bS)-1,1,9,9a-tetramethyl-3a,4,7,8,9,9b-hexahydronaphtho[2,1-c]dioxol-5-one
SMIL C[C@@H]1CCC=C2C(=O)C[C@H]3OOC(C)(C)[C@H]3[C@@]12C
Tekniske data
Kemisk navn eller materiale Nardosinone
Mængde 1 g
InChI formel InChI=1S/C15H22O3/c1-9-6-5-7-10-11(16)8-12-13(15(9,10)4)14(2,3)18-17-12/h7,9,12-13H,5-6,8H2,1-4H3/t9-,12-,13+,15+/m1/s1
Formel vægt 250.1569