missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Naratriptan Hydrochloride, LoGiCal
Specifications
Specifications
| Kemisk navn eller materiale | Naratriptan Hydrochloride |
| CAS | 143388-64-1 |
| Analyt- eller komponentnavne | Naratriptan Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H25 N3 O2 S . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Naratriptan Hydrochloride, 1H-Indole-5-ethanesulfonamide, N-methyl-3-(1-methyl-4-piperidinyl)-, hydrochloride (1:1), 1H-Indole-5-ethanesulfonamide, N-methyl-3-(1-methyl-4-piperidinyl)-, monohydrochloride (9CI), Amerge, GR 85548A, N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulfonamide hydrochloride, Naramig, Naratrex 1, GR 85548A, GR 85548A |
| InChI formel | InChI=1S/C17H25N3O2S.ClH/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;/h3-4,11-12,14,18-19H,5-10H2,1-2H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CNS(=O)(=O)CCc1ccc2[nH]cc(C3CCN(C)CC3)c2c1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?