Få mere at vide
Naratriptan-d3 Hydrochloride, TRC
Artikelnummer. 30482857
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30482857 10 mg
30472933 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482857 Leverandør Toronto Research Chemicals Leverandørnr. TRCN37873210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1190021-64-7
Molekylær formel C17H23D3ClN3O2S
Molekylvægt (g/mol) 374.94
Synonym N-Methyl-d3-2-[3-(1-methyl-4-piperidyl)-1H-indol-5-yl]-ethanesulfonamide Hydrochloride, Naramig-d3, Amerge-d3.
SMIL O=S(CCC1=CC=C2C(C(C3CCN(C([2H])([2H])[2H])CC3)=CN2)=C1)(NC)=O.Cl
Tekniske data
Kemisk navn eller materiale Naratriptan-d3 Hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C17H25N3O2S.ClH/c1-18-23(21,22)10-7-13-3-4-17-15(11-13)16(12-19-17)14-5-8-20(2)9-6-14;/h3-4,11-12,14,18-19H,5-10H2,1-2H3;1H/i2D3;
Formel vægt 374.1623