Learn More
Emma Safety Footwear Napoli Safety Shoes
Virtually impenetrable nitrile rubber soles combined with the shoe's sophisticated looks allow you to move easily between the office and warehouse.
Brand:  Emma Safety Footwear MM11109047
Additional Details : Weight : 1.79100kg
 View more versions of this product
Product Code. 15958181
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Nitrile Rubber Heat Resistant Outsole
  • Full grain leather/Nubuck
  • ESD Certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Brown
Hydro-Tec™ Sanitized Silver & Wellness leather
47
Laces
Napoli
Leather, Nubuck
Steel