Få mere at vide
Naphtol AS, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Napthol AS |
| CAS | 92-77-3 |
| Molekylær formel | C17 H13 N O2 |
| Mængde | 100 g |
| Synonym | 2-Naphthanilide','3-hydroxy- (8CI), 3-Hydroxy-N-phenyl-2-naphthalenecarboxamide, 2-Hydroxy-3-(phenylcarbamoyl)naphthalene, 2-Hydroxy-3-naphthalenecarboxanilide, 2-Hydroxy-3-naphthanilide, 2-Hydroxy-3-naphthoic acid anilide, 2-Hydroxy-3-naphthoic anilide, 2-Hydroxy-N-phenyl-3-naphthalenecarboxamide, 2-Naphthol-3-carboxylic acid N-phenylamide, 3-(N-Phenylcarbamoyl)-2-naphthol, 3-Hydroxy-2-naphthalenecarboxanilide, 3-Hydroxy-2-naphthanilide, 3-Hydroxy-2-naphthoanilide, 3-Hydroxy-2-naphthoic acid anilide, 3-Hydroxy-N-phenyl-2-naphthamide, 3-Hydroxyl-2-naphthanilide, Acco Naphthol AS, Acna Naphthol C, Amanil Naphthol AS, Amarthol AS, Anathol AS, Anthonaphthol AS, Aquafine Red E 9, Azoground AS, Azoic Coupling Component 2, Azonaphthol A, Azotol A, Brenthol AS, C.I. 37505, C.I. Azo Coupling Comp. 2, C.I. Azoic Coupling Component 2, Celcot RF, Cibanaphthol RF, Conazoic Coupling B, Diathol AS, Diathol ASF, Dragonthol A, Dycosthol AS, Hebeithol AS, Hiltonaphthol AS, Kako Grounder AS, Kambothol AS, Kiwa Grounder AS, Lake Developer A, Mitsui Naphthozol AS, N-Phenyl-2-hydroxy-3-naphthalenecarboxamide, N-Phenyl-3-hydroxy-2-naphthamide, NSC 45173, Naftol AS, Naftolo MM, Naphtanilide RC, Naphthanil AS, Naphthanilide OL Supra, Naphthanilide RC Supra, Naphthazol A, Naphthoide AS, Naphthoide AS No. 100, Naphthol ACNA C, Naphthol AS, Naphthol AS Supra, Naphtholate AS, Naphtoelan A, Naphtol AS, Naphtol AS-A, Napthol AS, Orange Base GC, Sefen AS, Solunaptol A, Tulathol AS, Ultrazol I-AS, α-Naphthol AS |
| InChI formel | InChI=1S/C17H13NO2/c19-16-11-13-7-5-4-6-12(13)10-15(16)17(20)18-14-8-2-1-3-9-14/h1-11,19H,(H,18,20) |
| SMIL | Oc1cc2ccccc2cc1C(=O)Nc3ccccc3 |
| IUPAC navn | 3-hydroxy-N-phenylnaphthalene-2-carboxamide |
| Molekylvægt (g/mol) | 263.29 |
| Formel vægt | 263.0946 |