missing translation for 'onlineSavingsMsg'
Få mere at vide

Naphtol AS, TRC

Artikelnummer. 30464953
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464953 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464953 Leverandør Toronto Research Chemicals Leverandørnr. TRCN4970605G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Napthol AS
CAS 92-77-3
Molekylær formel C17 H13 N O2
Mængde 5 g
Synonym 2-Naphthanilide','3-hydroxy- (8CI), 3-Hydroxy-N-phenyl-2-naphthalenecarboxamide, 2-Hydroxy-3-(phenylcarbamoyl)naphthalene, 2-Hydroxy-3-naphthalenecarboxanilide, 2-Hydroxy-3-naphthanilide, 2-Hydroxy-3-naphthoic acid anilide, 2-Hydroxy-3-naphthoic anilide, 2-Hydroxy-N-phenyl-3-naphthalenecarboxamide, 2-Naphthol-3-carboxylic acid N-phenylamide, 3-(N-Phenylcarbamoyl)-2-naphthol, 3-Hydroxy-2-naphthalenecarboxanilide, 3-Hydroxy-2-naphthanilide, 3-Hydroxy-2-naphthoanilide, 3-Hydroxy-2-naphthoic acid anilide, 3-Hydroxy-N-phenyl-2-naphthamide, 3-Hydroxyl-2-naphthanilide, Acco Naphthol AS, Acna Naphthol C, Amanil Naphthol AS, Amarthol AS, Anathol AS, Anthonaphthol AS, Aquafine Red E 9, Azoground AS, Azoic Coupling Component 2, Azonaphthol A, Azotol A, Brenthol AS, C.I. 37505, C.I. Azo Coupling Comp. 2, C.I. Azoic Coupling Component 2, Celcot RF, Cibanaphthol RF, Conazoic Coupling B, Diathol AS, Diathol ASF, Dragonthol A, Dycosthol AS, Hebeithol AS, Hiltonaphthol AS, Kako Grounder AS, Kambothol AS, Kiwa Grounder AS, Lake Developer A, Mitsui Naphthozol AS, N-Phenyl-2-hydroxy-3-naphthalenecarboxamide, N-Phenyl-3-hydroxy-2-naphthamide, NSC 45173, Naftol AS, Naftolo MM, Naphtanilide RC, Naphthanil AS, Naphthanilide OL Supra, Naphthanilide RC Supra, Naphthazol A, Naphthoide AS, Naphthoide AS No. 100, Naphthol ACNA C, Naphthol AS, Naphthol AS Supra, Naphtholate AS, Naphtoelan A, Naphtol AS, Naphtol AS-A, Napthol AS, Orange Base GC, Sefen AS, Solunaptol A, Tulathol AS, Ultrazol I-AS, α-Naphthol AS
InChI formel InChI=1S/C17H13NO2/c19-16-11-13-7-5-4-6-12(13)10-15(16)17(20)18-14-8-2-1-3-9-14/h1-11,19H,(H,18,20)
SMIL Oc1cc2ccccc2cc1C(=O)Nc3ccccc3
IUPAC navn 3-hydroxy-N-phenylnaphthalene-2-carboxamide
Molekylvægt (g/mol) 263.29
Formel vægt 263.0946
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.